siRNA Details
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047877.1 | NP_001041342.1 | 6 | 760 | ||
XM_006247599.2 | XP_006247661.1 | 8 | 2960 | |||
XM_006247600.2 | XP_006247662.1 | 7 | 2817 | |||
XM_006247601.2 | XP_006247663.1 | 9 | 3186 | |||
XM_006247602.2 | XP_006247664.1 | 8 | 1748 | |||
XM_006247603.2 | XP_006247665.1 | 8 | 1602 | |||
XM_006247604.2 | XP_006247666.1 | 8 | 3018 | |||
XM_006247606.2 | XP_006247668.1 | 7 | 2875 | |||
XM_006247607.2 | XP_006247669.1 | 7 | 1787 | |||
XM_006247608.2 | XP_006247670.1 | 6 | 1437 | |||
XM_006247609.2 | XP_006247671.1 | 6 | 732 | |||
XM_008768368.1 | XP_008766590.1 | 9 | 3129 | |||
XM_008768369.1 | XP_008766591.1 | 10 | 2593 |
