siRNA Details
Gene Symbol: Trim65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135714.1 | NP_001129186.1 | 3 | 1067 | ||
GenBank mRNA | FQ223610.1 | - | 638 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135714.1 | NP_001129186.1 | 3 | 1067 | ||
GenBank mRNA | FQ223610.1 | - | 638 |