siRNA Details
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198760.1 | NP_942055.1 | 6 | 1462 | ||
XM_006247613.2 | XP_006247675.1 | 7 | 4140 | |||
XM_006247614.2 | XP_006247676.1 | 7 | 2161 | |||
XM_006247617.2 | XP_006247679.1 | 8 | 1845 | |||
XM_006247618.2 | XP_006247680.1 | 7 | 1727 | |||
XM_006247619.2 | XP_006247681.1 | 6 | 3683 | |||
XM_006247620.2 | XP_006247682.1 | 6 | 3364 | |||
XM_008768375.1 | XP_008766597.1 | 6 | 1973 | |||
XM_008768376.1 | XP_008766598.1 | 7 | 3871 | |||
GenBank mRNA | AY325107.1 | - | 6 | 1462 | ||
BC081999.1 | - | 6 | 1523 | 7131044 | ||
FQ212616.1 | - | 2 | 798 |
