siRNA Details
Gene Symbol: Aifm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768872.1 | XP_008767094.1 | 4 | 2563 | ||
XM_008768873.1 | XP_008767095.1 | 4 | 2563 | |||
XM_008768874.1 | XP_008767096.1 | 4 | 2563 | |||
XM_008768875.1 | XP_008767097.1 | 5 | 2087 | |||
XM_008768876.1 | 5 | 2087 |
