siRNA Details
Gene Symbol: Aifm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768872.1 | XP_008767094.1 | 8, 9 | 2980 | ||
XM_008768873.1 | XP_008767095.1 | 8, 9 | 2980 | |||
XM_008768874.1 | XP_008767096.1 | 8, 9 | 2980 | |||
XM_008768875.1 | XP_008767097.1 | 9, 10 | 2504 | |||
XM_008768876.1 | 9, 10 | 2504 | ||||
GenBank mRNA | BC089949.1 | - | 1, 2 | 291 |

More Information
Other Species Targeted

Gene Ontology Categories:
