siRNA Details
Gene Symbol: Parp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103351.2 | NP_001096821.1 | 8 | 1905 | ||
XM_006248411.1 | XP_006248473.1 | 8 | 1731 | |||
XM_008768727.1 | XP_008766949.1 | 8 | 3014 | |||
XM_008768728.1 | XP_008766950.1 | 7 | 3659 | |||
XM_008768729.1 | XP_008766951.1 | 8 | 1835 | |||
GenBank mRNA | BC128756.1 | - | 8 | 1908 | 7105391 |
