siRNA Details
Gene Symbol: Col8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107100.1 | NP_001100570.1 | 4 | 2417 | ||
XM_006248207.1 | XP_006248269.1 | 2 | 2226 | |||
XM_006248208.2 | XP_006248270.1 | 2 | 2225 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007739, AK032048, AK018742, BC011061, AK046370, and others.
Gene Ontology Categories:
