siRNA Details
Gene Symbol: Ttyh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107124.1 | NP_001100594.1 | 5 | 668 | ||
XM_008768969.1 | XP_008767191.1 | 5 | 857 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttyh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107124.1 | NP_001100594.1 | 5 | 668 | ||
XM_008768969.1 | XP_008767191.1 | 5 | 857 |