siRNA Details
Gene Symbol: Ap4m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037977.1 | NP_001033066.1 | 10 | 787 | ||
XM_006249008.2 | XP_006249070.1 | 10 | 816 | |||
GenBank mRNA | BC127443.1 | - | 10 | 852 | ||
BC161814.1 | - | 11 | 981 | |||
DQ298439.1 | - | 10 | 787 |
