siRNA Details
Gene Symbol: RGD1566138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769159.1 | XP_008767381.1 | 5 | 675 |
Gene Symbol: LOC100910636 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751148.3 | XP_003751196.1 | 5 | 671 | ||
XM_006249080.2 | XP_006249142.1 | 5 | 669 | |||
XM_008769105.1 | XP_008767327.1 | 5 | 671 |
Gene Symbol: Zcwpw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249069.2 | XP_006249131.1 | 4 | 828 |
