siRNA Details
Gene Symbol: Git2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005553.1 | NP_001005553.1 | 4 | 427 | ||
XM_006249442.2 | XP_006249504.1 | 4 | 475 | |||
XM_006249445.2 | XP_006249507.1 | 4 | 475 | |||
XM_006249447.2 | XP_006249509.1 | 4 | 475 | |||
XM_006249450.2 | XP_006249512.1 | 4 | 475 | |||
XM_008769324.1 | XP_008767546.1 | 4 | 475 | |||
XM_008769325.1 | XP_008767547.1 | 4 | 475 | |||
GenBank mRNA | BC081967.1 | - | 4 | 427 | 7125406 |
