siRNA Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107157.1 | NP_001100627.1 | 7 | 1571 | ||
XM_006249158.1 | XP_006249220.1 | 6 | 1517 | |||
XM_008769137.1 | XP_008767359.1 | 6 | 1372 | |||
GenBank mRNA | BC166738.1 | - | 6 | 1458 | 7937292 |

More Information
Other Species Targeted

Gene Ontology Categories:
