siRNA Details
Gene Symbol: Rrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135846.1 | NP_001129318.1 | 14 | 1325 | ||
XM_008767458.1 | XP_008765680.1 | 14 | 1349 | |||
XM_008767459.1 | XP_008765681.1 | 14 | 1700 | |||
GenBank mRNA | BC166485.1 | - | 14 | 1325 | 7099831 | |
FQ228986.1 | - | 14 | 1345 |
