siRNA Details
Gene Symbol: Aim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250335.1 | XP_006250397.1 | 4 | 945 | ||
XM_006250336.2 | XP_006250398.1 | 5 | 1389 | |||
XM_006250339.1 | XP_006250401.1 | 3 | 760 | |||
XM_008769802.1 | XP_008768024.1 | 5 | 2604 | |||
XM_008769803.1 | XP_008768025.1 | 5 | 3554 | |||
XM_008769804.1 | XP_008768026.1 | 6 | 2932 | |||
XM_008769805.1 | XP_008768027.1 | 5 | 2541 | |||
XM_222949.8 | XP_222949.5 | 4 | 3369 |
