siRNA Details
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107199.1 | NP_001100669.1 | 15 | 1699 | ||
GenBank mRNA | AY724501.1 | - | 15 | 1688 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107199.1 | NP_001100669.1 | 15 | 1699 | ||
GenBank mRNA | AY724501.1 | - | 15 | 1688 |