siRNA Details
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107199.1 | NP_001100669.1 | 3, 4 | 388 | ||
GenBank mRNA | AY724501.1 | - | 3, 4 | 377 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172650, AK139700, AK034380, BC061458, BC079581, and others.
Gene Ontology Categories:
