siRNA Details
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107200.1 | NP_001100670.1 | 12 | 2139 | ||
XM_006250453.2 | XP_006250515.1 | 13 | 2309 | |||
XM_008769853.1 | XP_008768075.1 | 12 | 1927 | |||
XM_008769854.1 | XP_008768076.1 | 13 | 1959 |
