siRNA Details
Gene Symbol: Lrrc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107204.2 | NP_001100674.1 | 1 | 2123 | ||
XM_006250557.1 | XP_006250619.1 | 4 | 2434 | |||
GenBank mRNA | FQ212117.1 | - | 4 | 2439 | ||
FQ217980.1 | - | 4 | 2389 |
