siRNA Details
Gene Symbol: Hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009684.1 | NP_001009684.1 | 1 | 221 | ||
GenBank mRNA | BC088191.1 | - | 1 | 221 | 7372031 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009684.1 | NP_001009684.1 | 1 | 221 | ||
GenBank mRNA | BC088191.1 | - | 1 | 221 | 7372031 |