siRNA Details
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013989.1 | NP_001014011.1 | 4 | 596 | ||
XM_006250733.2 | XP_006250795.1 | 4 | 590 | |||
XM_006250734.2 | XP_006250796.1 | 4 | 645 | |||
XM_006250735.1 | XP_006250797.1 | 4 | 428 | |||
XM_006250736.2 | XP_006250798.1 | 3 | 239 | |||
GenBank mRNA | BC079225.1 | - | 4 | 596 | 7132309 | |
FQ212261.1 | - | 4 | 433 |
