siRNA Details
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107222.1 | NP_001100692.1 | 10 | 1900 | ||
XM_006251076.2 | XP_006251138.1 | 11 | 2089 | |||
GenBank mRNA | FQ212545.1 | - | 3 | 561 |
