siRNA Details
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107226.1 | NP_001100696.1 | 1 | 131 | ||
XM_006251125.2 | XP_006251187.1 | 2 | 662 | |||
XM_006251126.2 | XP_006251188.1 | 3 | 473 | |||
XM_006251127.2 | XP_006251189.1 | 2 | 274 | |||
XM_006251128.2 | XP_006251190.1 | 2 | 1787 | |||
XM_006251130.2 | XP_006251192.1 | 2 | 661 | |||
XM_008770281.1 | XP_008768503.1 | 2 | 924 | |||
XM_008770282.1 | XP_008768504.1 | 3 | 348 |
