siRNA Details
Gene Symbol: Prr14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251416.2 | XP_006251478.1 | 5 | 4525 | ||
XM_008770385.1 | XP_008768607.1 | 5 | 4529 | |||
XM_008770386.1 | XP_008768608.1 | 5 | 5239 | |||
XM_008770387.1 | XP_008768609.1 | 5 | 4855 | |||
XM_008770388.1 | XP_008768610.1 | 4 | 4718 | |||
XM_223586.7 | XP_223586.5 | 4 | 4366 |
