siRNA Details
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107236.1 | NP_001100706.1 | 8 | 2231 | ||
XM_008770308.1 | XP_008768530.1 | 10 | 3917 | |||
XM_008770309.1 | XP_008768531.1 | 9 | 3842 | |||
XM_008770311.1 | XP_008768533.1 | 10 | 4014 |
