siRNA Details
Gene Symbol: Ccm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126275.1 | NP_001119747.1 | 2, 3 | 240 | ||
XM_006251318.2 | XP_006251380.1 | 2, 3 | 2084 | |||
XM_006251319.2 | XP_006251381.1 | 2, 3 | 525 | |||
GenBank mRNA | BC160889.1 | - | 2, 3 | 240 | 7441909 |
