siRNA Details
Gene Symbol: Zfp385d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013992.1 | NP_001014014.1 | 2 | 416 | ||
XM_008770513.1 | XP_008768735.1 | 1 | 3248 | |||
GenBank mRNA | BC079278.1 | - | 2 | 416 | 7134172 |
