siRNA Details
Gene Symbol: Kctd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107253.1 | NP_001100723.1 | 2 | 492 | ||
XM_006251763.2 | XP_006251825.1 | 3 | 2127 | |||
XM_006251765.2 | XP_006251827.1 | 2 | 3029 | |||
GenBank mRNA | BC158799.1 | - | 1 | 438 |
