siRNA Details
Gene Symbol: Rpgrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251921.2 | XP_006251983.1 | 14 | 1743 | ||
XM_008770631.1 | XP_008768853.1 | 14 | 1711 | |||
XM_008770632.1 | XP_008768854.1 | 14 | 1741 | |||
XM_008770633.1 | XP_008768855.1 | 13 | 1613 | |||
XM_008770634.1 | XP_008768856.1 | 14 | 1740 | |||
XM_008770635.1 | XP_008768857.1 | 14 | 1736 | |||
XM_008770636.1 | XP_008768858.1 | 14 | 1736 | |||
GenBank mRNA | BC097397.1 | - | 4 | 653 |
