siRNA Details
Gene Symbol: Sall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107262.1 | NP_001100732.1 | 2 | 3512 | ||
XM_006251884.2 | XP_006251946.1 | 2 | 3047 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107262.1 | NP_001100732.1 | 2 | 3512 | ||
XM_006251884.2 | XP_006251946.1 | 2 | 3047 |