siRNA Details
Gene Symbol: Zfhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098803.1 | NP_001092273.1 | 9 | 5135 | ||
XM_008770677.1 | XP_008768899.1 | 8 | 6910 | |||
XM_008770678.1 | XP_008768900.1 | 7 | 6835 | |||
GenBank mRNA | AY237644.1 | - | 1 | 445 |
