siRNA Details
Gene Symbol: Rnf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191685.1 | NP_001178614.1 | 12, 13 | 1471 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191685.1 | NP_001178614.1 | 12, 13 | 1471 |