siRNA Details
Gene Symbol: Dis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127483.1 | NP_001120955.1 | 16 | 2180 | ||
XM_006252391.2 | XP_006252453.1 | 17 | 2041 | |||
XM_008770869.1 | XP_008769091.1 | 16 | 2115 | |||
GenBank mRNA | BC166824.1 | - | 16 | 2180 | 8370691 | |
FQ210067.1 | - | 16 | 2192 |
