siRNA Details
Gene Symbol: Spry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012046.1 | NP_001012046.1 | 2 | 319 | ||
XM_006252438.2 | XP_006252500.1 | 2 | 613 | |||
GenBank mRNA | BC089116.1 | - | 2 | 319 | 7320532 |

More Information
Other Species Targeted

Gene Ontology Categories:
