siRNA Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100685.1 | NP_001094155.1 | 6, 7 | 804 | ||
XM_006252508.2 | XP_006252570.1 | 5, 6 | 633 | |||
GenBank mRNA | BC088243.1 | - | 6, 7 | 784 | ||
FQ214276.1 | - | 5, 6 | 589 |
