siRNA Details
Gene Symbol: Il17rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107290.1 | NP_001100760.1 | 11 | 1252 | ||
XM_006252603.2 | XP_006252665.1 | 11 | 1234 | |||
GenBank mRNA | BC168886.1 | - | 7 | 797 | 7369142 |
