siRNA Details
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013134.2 | NP_001013152.2 | 11 | 1958 | ||
XM_008770996.1 | XP_008769218.1 | 11 | 2062 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013134.2 | NP_001013152.2 | 11 | 1958 | ||
XM_008770996.1 | XP_008769218.1 | 11 | 2062 |