siRNA Details
Gene Symbol: Lonrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771313.1 | XP_008769535.1 | 6 | 1410 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lonrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771313.1 | XP_008769535.1 | 6 | 1410 |