siRNA Details
Gene Symbol: Got1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014003.1 | NP_001014025.1 | 5 | 629 | ||
XM_006253291.2 | XP_006253353.1 | 5 | 1000 | |||
XM_006253292.2 | XP_006253354.1 | 5 | 520 | |||
XM_006253294.2 | XP_006253356.1 | 5 | 1000 | |||
XM_008771326.1 | XP_008769548.1 | 5 | 662 | |||
XM_008771327.1 | XP_008769549.1 | 5 | 1000 | |||
XM_008771328.1 | XP_008769550.1 | 5 | 1000 | |||
GenBank mRNA | BC079187.1 | - | 5 | 629 | 7131193 |
