siRNA Details
Gene Symbol: Mcph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253383.2 | XP_006253445.1 | 7 | 1447 | ||
XM_006253385.2 | XP_006253447.1 | 7 | 800 | |||
XM_006253388.2 | XP_006253450.1 | 7 | 1445 | |||
XM_225006.8 | XP_225006.7 | 7 | 1448 |
