siRNA Details
Gene Symbol: Tubgcp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107323.1 | NP_001100793.1 | 7 | 809 | ||
XM_006253440.2 | XP_006253502.1 | 7 | 837 | |||
XM_006253441.2 | XP_006253503.1 | 6 | 729 | |||
GenBank mRNA | FQ222809.1 | - | 9 | 1031 |
