siRNA Details
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107324.2 | NP_001100794.2 | 25, 26 | 3294 | ||
XM_006253446.2 | XP_006253508.1 | 25, 26 | 3249 | |||
XM_006253447.2 | XP_006253509.1 | 25, 26 | 3249 | |||
XM_006253448.2 | XP_006253510.1 | 25, 26 | 3249 |
