siRNA Details
Gene Symbol: Papd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107333.1 | NP_001100803.1 | 2 | 242 | ||
XM_006227781.2 | XP_006227843.1 | 2 | 3992 | |||
XM_006227782.2 | XP_006227844.1 | 2 | 629 | |||
XM_008758690.1 | XP_008756912.1 | 2 | 359 |
