siRNA Details
Gene Symbol: LOC102549404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771480.1 | XP_008769702.1 | 4 | 525 |
Gene Symbol: Spock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271297.1 | NP_001258226.1 | 264 | |||
GenBank mRNA | CB608950.1 | - | 273 |
