siRNA Details
Gene Symbol: Susd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107341.1 | NP_001100811.1 | 5 | 667 | ||
XM_006253709.2 | XP_006253771.1 | 5 | 2243 | |||
GenBank mRNA | FQ227785.1 | - | 5 | 852 |
