siRNA Details
Gene Symbol: Ankrd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033698.1 | NP_001028870.1 | 2, 3 | 616 | ||
GenBank mRNA | BC099837.1 | - | 2, 3 | 616 | 7936269 |

More Information
Other Species Targeted

