siRNA Details
Gene Symbol: Mcm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107366.1 | NP_001100836.1 | 4 | 529 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mcm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107366.1 | NP_001100836.1 | 4 | 529 |