siRNA Details
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107396.1 | NP_001100866.1 | 9 | 1274 | ||
XM_006254579.2 | XP_006254641.1 | 9 | 1192 | |||
XM_006254580.2 | XP_006254642.1 | 9 | 1192 | |||
XM_008772039.1 | XP_008770261.1 | 10 | 1104 |
