siRNA Details
Gene Symbol: Epb41l4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107397.2 | NP_001100867.1 | 8 | 706 | ||
XM_006254582.2 | XP_006254644.1 | 13 | 3978 | |||
XM_006254583.2 | XP_006254645.1 | 13 | 3084 | |||
XM_006254584.2 | XP_006254646.1 | 14 | 2730 | |||
XM_006254588.2 | XP_006254650.1 | 13 | 1284 | |||
XM_008772040.1 | XP_008770262.1 | 13 | 1061 | |||
GenBank mRNA | BC168858.1 | - | 8 | 706 | 7097125 |

More Information
Other Species Targeted

