siRNA Details
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107399.1 | NP_001100869.1 | 6 | 912 | ||
XM_006254590.2 | XP_006254652.1 | 6 | 1726 | |||
XM_006254591.2 | XP_006254653.1 | 6 | 1763 | |||
XM_006254593.2 | XP_006254655.1 | 6 | 1163 | |||
XM_008772042.1 | XP_008770264.1 | 6 | 1763 |
