siRNA Details
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772250.1 | XP_008770472.1 | 8 | 938 | ||
XM_008772251.1 | XP_008770473.1 | 8 | 859 | |||
XM_008772252.1 | XP_008770474.1 | 8 | 1149 | |||
XM_008772253.1 | XP_008770475.1 | 7 | 1017 | |||
XM_008772254.1 | XP_008770476.1 | 8 | 1149 | |||
XM_008772257.1 | XP_008770479.1 | 8 | 1149 |
